Merck
  • Synthesis of isoxazoles en route to semi-aromatized polyketides: dehydrogenation of benzonitrile oxide-para-quinone acetal cycloadducts.
Synthesis of isoxazoles en route to semi-aromatized polyketides: dehydrogenation of benzonitrile oxide-para-quinone acetal cycloadducts.
Organic & biomolecular chemistry (2012-05-19)
Yoshimitsu Hashimoto, Akiomi Takada, Hiroshi Takikawa, Keisuke Suzuki
ABSTRACT
A variety of highly functionalized polycyclic isoxazoles are prepared by a two-step protocol: (1) 1,3-dipolar cycloaddition of o,o'-disubstituted benzonitrile oxides to para-quinone mono-acetals, then (2) dehydrogenation. The cycloaddition proceeds in a regioselective manner, favouring the formation of the 4-acyl cycloadducts, which are suitable intermediates for the synthesis of semi-aromatized polycyclic targets derived from polyketide type-II biosynthesis.
Product Description
Supelco
Benzonitrile, suitable for HPLC, 99.9%
Sigma-Aldrich
Benzonitrile, ReagentPlus®, 99%