Merck
  • The in vitro and in vivo genotoxicity of benzocaine: a brief communication.
The in vitro and in vivo genotoxicity of benzocaine: a brief communication.
International journal of toxicology (2012-05-05)
William J Brock, Thomas A Bell
ABSTRACT
Benzocaine has a long history of use in human medicine. However, benzocaine also has been used in aquaculture with finfish for more than 40 years for sedating fish for marking, transport, surgery, and so on, although benzocaine does not have a current Food and Drug Administration (FDA) approval for this application in the United States. As part of a FDA approval for use as an animal drug, the genotoxicity of benzocaine was evaluated in the in vitro bacterial reverse mutation assay and the forward mutation assay and in vivo in the mouse micronucleus assay. These studies were conducted in compliance with Good Laboratory Practice regulations and according to Veterinary International Conference on Harmonisation guidelines. Based on the results of these studies, benzocaine was determined not to be genotoxic.
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)