Merck
  • Characterization of the modes of binding between human sweet taste receptor and low-molecular-weight sweet compounds.
Characterization of the modes of binding between human sweet taste receptor and low-molecular-weight sweet compounds.
PloS one (2012-04-27)
Katsuyoshi Masuda, Ayako Koizumi, Ken-ichiro Nakajima, Takaharu Tanaka, Keiko Abe, Takumi Misaka, Masaji Ishiguro
ABSTRACT
One of the most distinctive features of human sweet taste perception is its broad tuning to chemically diverse compounds ranging from low-molecular-weight sweeteners to sweet-tasting proteins. Many reports suggest that the human sweet taste receptor (hT1R2-hT1R3), a heteromeric complex composed of T1R2 and T1R3 subunits belonging to the class C G protein-coupled receptor family, has multiple binding sites for these sweeteners. However, it remains unclear how the same receptor recognizes such diverse structures. Here we aim to characterize the modes of binding between hT1R2-hT1R3 and low-molecular-weight sweet compounds by functional analysis of a series of site-directed mutants and by molecular modeling-based docking simulation at the binding pocket formed on the large extracellular amino-terminal domain (ATD) of hT1R2. We successfully determined the amino acid residues responsible for binding to sweeteners in the cleft of hT1R2 ATD. Our results suggest that individual ligands have sets of specific residues for binding in correspondence with the chemical structures and other residues responsible for interacting with multiple ligands.
Product Description
Supelco
Aspartame, analytical standard
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
Aspartame, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Supelco
Sucralose, analytical standard