Merck
  • Achiral counterion control of enantioselectivity in a Brønsted acid-catalyzed iodolactonization.
Achiral counterion control of enantioselectivity in a Brønsted acid-catalyzed iodolactonization.
Journal of the American Chemical Society (2012-04-03)
Mark C Dobish, Jeffrey N Johnston
ABSTRACT
Highly enantioselective halolactonizations have been developed that employ a chiral proton catalyst-N-iodosuccinimide (NIS) reagent system in which the Brønsted acid is used at catalyst loadings as low as 1 mol %. An approach that modulates the achiral counterion (equimolar to the neutral chiral ligand-proton complex present at low catalyst loadings) to optimize the enantioselection is documented for the first time in this transformation. In this way, unsaturated carboxylic acids are converted to γ-lactones in high yields (up to 98% ee) using commercially available NIS.
Sigma-Aldrich
N-Iodosuccinimide, 95%