Merck
  • Sumatriptan (rectal route of administration) for acute migraine attacks in adults.
Sumatriptan (rectal route of administration) for acute migraine attacks in adults.
The Cochrane database of systematic reviews (2012-02-18)
Christopher J Derry, Sheena Derry, R Andrew Moore
ABSTRACT
Migraine is a highly disabling condition for the individual and also has wide-reaching implications for society, healthcare services, and the economy. Sumatriptan is an abortive medication for migraine attacks, belonging to the triptan family. Rectal administration may be preferable to oral for individuals experiencing nausea and/or vomiting. To determine the efficacy and tolerability of rectal sumatriptan compared to placebo and other active interventions in the treatment of acute migraine attacks in adults. We searched the Cochrane Central Register of Controlled Trials (CENTRAL), MEDLINE, EMBASE, online databases, and reference lists for studies through 13 October 2011. We included randomised, double-blind, placebo- and/or active-controlled studies using rectally administered sumatriptan to treat a migraine headache episode, with at least 10 participants per treatment arm. Two review authors independently assessed trial quality and extracted data. We used numbers of participants achieving each outcome to calculate relative risk (or 'risk ratio') and numbers needed to treat to benefit (NNT) or harm (NNH) compared to placebo or a different active treatment. Three studies (866 participants) compared rectally administered sumatriptan with placebo or an active comparator. Most of the data were for the 12.5 mg and 25 mg doses. For the majority of efficacy outcomes, sumatriptan surpassed placebo. For sumatriptan 12.5 mg versus placebo the NNTs were 5.2 and 3.2 for headache relief at one and two hours, respectively. Results for the 25 mg dose were similar to the 12.5 mg dose, and there were no significant differences between the two doses for any of the outcomes analysed. The NNTs for sumatriptan 25 mg versus placebo were 4.2, 3.2, and 2.4 for pain-free at two hours, headache relief at one hour, and headache relief at two hours, respectively.Relief of functional disability was greater with sumatriptan than with placebo, with NNTs of 8.0 and 4.0 for the 12.5 mg and 25 mg doses, respectively. For the most part, adverse events were transient and mild and were more common with sumatriptan than with placebo, but there were insufficient data to perform any analyses.Direct comparison of sumatriptan with active treatments was limited to one study comparing sumatriptan 25 mg with ergotamine tartrate 2 mg + caffeine 100 mg. Based on limited amounts of data, sumatriptan 25 mg, administered rectally, is an effective treatment for acute migraine attacks, with participants in these studies experiencing a significant reduction in headache pain and functional disability within two hours of treatment. The lack of data on relief of headache-associated symptoms or incidence of adverse events limits any conclusions that can be drawn.
Sigma-Aldrich
Ergotamine D-tartrate, ≥97.0% (calc. based on dry substance, NT)
Pricing and availability is not currently available.