Merck
  • Overexpression of long non-coding RNAs following exposure to xenobiotics in the aquatic midge Chironomus riparius.
Overexpression of long non-coding RNAs following exposure to xenobiotics in the aquatic midge Chironomus riparius.
Aquatic toxicology (Amsterdam, Netherlands) (2012-01-27)
José-Luis Martínez-Guitarte, Rosario Planelló, Gloria Morcillo
ABSTRACT
Non-coding RNAs (ncRNAs) represent an important transcriptional output of eukaryotic genomes. In addition to their functional relevance as housekeeping and regulatory elements, recent studies have suggested their involvement in rather unexpected cellular functions. The aim of this work was to analyse the transcriptional behaviour of non-coding RNAs in the toxic response to pollutants in Chironomus riparius, a reference organism in aquatic toxicology. Three well-characterized long non-coding sequences were studied: telomeric repeats, Cla repetitive elements and the SINE CTRT1. Transcription levels were evaluated by RT-PCR after 24-h exposures to three current aquatic contaminants: bisphenol A (BPA), benzyl butyl phthalate (BBP) and the heavy metal cadmium (Cd). Upregulation of telomeric transcripts was found after BPA treatments. Moreover, BPA significantly activated Cla transcription, which also appeared to be increased by cadmium, whereas BBP did not affect the transcription levels of these sequences. Transcription of SINE CTRT1 was not altered by any of the chemicals tested. These data are discussed in the light of previous studies that have shown a response by long ncRNAS (lncRNAs) to cellular stressors, indicating a relationship with environmental stimuli. Our results demonstrated for the first time the ability of bisphenol A to activate non-coding sequences mainly located at telomeres and centromeres. Overall, this study provides evidence that xenobiotics can induce specific responses in ncRNAs derived from repetitive sequences that could be relevant in the toxic response, and also suggests that ncRNAs could represent a novel class of potential biomarkers in toxicological assessment.
Product Description
Supelco
Benzyl butyl phthalate, analytical standard
Sigma-Aldrich
Benzyl butyl phthalate, 98%