Merck
  • An approach to an inhibition electronic tongue to detect on-line organophosphorus insecticides using a computer controlled multi-commuted flow system.
An approach to an inhibition electronic tongue to detect on-line organophosphorus insecticides using a computer controlled multi-commuted flow system.
Sensors (Basel, Switzerland) (2011-12-14)
Gustavo A Alonso, Rocio B Dominguez, Jean-Louis Marty, Roberto Muñoz
ABSTRACT
An approach to an inhibition bioelectronic tongue is presented. The work is focused on development of an automated flow system to carry out experimental assays, a custom potentiostat to measure the response from an enzymatic biosensor, and an inhibition protocol which allows on-line detections. A Multi-commuted Flow Analysis system (MCFA) was selected and developed to carry out assays with an improved inhibition method to detect the insecticides chlorpyrifos oxon (CPO), chlorfenvinfos (CFV) and azinphos methyl-oxon (AZMO). The system manifold comprised a peristaltic pump, a set of seven electronic valves controlled by a personal computer electronic interface and software based on LabView® to control the sample dilutions into the cell. The inhibition method consists in the injection of the insecticide when the enzyme activity has reached the plateau of the current; with this method the incubation time is avoided. A potentiostat was developed to measure the response from the enzymatic biosensor. Low limits of detection of 10 nM for CPO, CFV, and AZMO were achieved.
Product Description
Supelco
Azinphos-methyl, PESTANAL®, analytical standard
Supelco
Chlorfenvinphos, PESTANAL®, analytical standard