MilliporeSigma
  • Ca2(+)-dependent interactions between the C-helix of troponin-C and troponin-I. Photocross-linking and fluorescence studies using a recombinant troponin-C.
Ca2(+)-dependent interactions between the C-helix of troponin-C and troponin-I. Photocross-linking and fluorescence studies using a recombinant troponin-C.
The Journal of biological chemistry (1990-03-25)
Z Y Wang, S Sarkar, J Gergely, T Tao
ABSTRACT
We have used in vitro mutagenesis to synthesize in Escherichia coli a recombinant rabbit skeletal troponin-C (designated as TnC57) in which Cys-98 was replaced with leucine, and Ala-57 in the C-helix of the N-terminal domain was replaced with cysteine. TnC57 labeled with the bifunctional photocross-linker benzophenone-4-maleimide could be photocross-linked with troponin-I in both the binary complex with troponin-I and in the ternary complex with troponin-I and troponin-T. The fluorescence lifetime of TnC57 labeled with the probe N-iodoacetyl-N'-(5-sulfo-1-naphthyl)ethylenediamine decreased from 13.2 +/- 0.1 to 11.8 +/- 0.1 ns when Ca2+ bound to the low affinity triggering sites. Complexation with either troponin-I or both troponin-I and troponin-T resulted in significant increases in this lifetime both in the absence and the presence of Ca2+. In either the binary or the ternary complex, this lifetime increased from 15.5 to 18.0 ns upon Ca2+ binding to the low affinity sites. Complementary acrylamide-quenching studies yielded results that are consistent with the fluorescence lifetime results. Our results show that the C-helix of troponin-C interacts with troponin-I, in confirmation of recent zero-length cross-linking results (Leszyk, J., Grabarek, Z., Gergely, J., and Collins, J.H. (1990) Biochemistry 29, 299-304). Moreover, they are in support of a model (Herzberg, O., Moult, J., and James, M.N.G. (1986) J. Biol. Chem. 261, 2638-2644) in which the binding of Ca2+ to the triggering sites in the N-terminal domain of troponin-C results in the movement of the B- and C-helices away from the central helix, thereby exposing a putative hydrophobic binding site for troponin-I.