Merck
  • Mutagenicity and genotoxicity of tannery effluents used for irrigation at Kanpur, India.
Mutagenicity and genotoxicity of tannery effluents used for irrigation at Kanpur, India.
Ecotoxicology and environmental safety (2010-08-06)
Mohammad Zubair Alam, Shamim Ahmad, Abdul Malik, Masood Ahmad
ABSTRACT
The tannery effluents at Kanpur (India) have been in use for irrigation since last many years, polluting soil directly while ground water and food crops indirectly. Gas chromatography-mass spectrometric analysis of the test samples revealed the presence of organic compounds including diisooctyl phthalate, phenyl N-methylcarbamate, dibutyl phthalate, bis 2-methoxyethyl phthalate, and higher alkanes. Tannery effluent extracts were prepared using XAD-4/8 resins, dichloromethane, chloroform, and hexane and tested with Ames Salmonella test and DNA repair-defective Escherichia coli K-12 mutants. In the presence of XAD-concentrated tannery effluent, TA98 found to be the most sensitive strain in terms of mutagenic index followed by TA97a whereas in terms of mutagenic potential TA102 was most responsive. The extracts were also found genotoxic as determined in terms of survival of E. coli K-12 mutants, suggesting the presence of DNA damaging compounds in the tannery effluents. In the light of results, precautious use of tannery effluents for irrigation is suggested.
Product Description
Supelco
Amberlite XAD4, 20-60 mesh
Pricing and availability is not currently available.
Supelco
Amberlite XAD4, 20-60 mesh
Pricing and availability is not currently available.
Supelco
Amberlite XAD4
Pricing and availability is not currently available.
Supelco
Amberlite XAD4, 20-60 mesh
Pricing and availability is not currently available.