Merck
  • Effect of PMMA polymer on the dynamic viscoelasticity and plasticizer leachability of PEMA-based tissue conditioners.
Effect of PMMA polymer on the dynamic viscoelasticity and plasticizer leachability of PEMA-based tissue conditioners.
Dental materials journal (2010-07-09)
Guang Hong, Takeshi Maeda, YingAi Li, Shinsuke Sadamori, Taizo Hamada, Hiroshi Murata
ABSTRACT
The purpose of this study was to determine the effect of PMMA polymer on dynamic viscoelasticity and plasticizer leachability of PEMA-based tissue conditioners. One PEMA polymer and one PMMA polymer were used in powder form with four formulations. The combination of 80 wt% ATBC, 15 wt% BPBG and 5 wt% ethyl alcohol was used as the liquid phase. The dynamic viscoelasticity and plasticizer leaching of each specimen were measured after 0, 1, 3, 7, and 14 days of immersion (37 degrees C distilled water) using DMA and HPLC. A significant difference was found among the materials in the dynamic viscoelasticity and leaching of plasticizer. The materials containing 10 wt% PMMA showed the most stable dynamic viscoelasticity, and showed the lowest leaching of plasticizer. The results suggest that the addition of the PMMA polymer to the powder of a tissue conditioner can improve the durability of the PEMA-based tissue conditioner.
Product Description
Supelco
Tributyl 2-acetylcitrate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Tributyl 2-acetylcitrate, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Tributyl O-acetylcitrate, 98%
Pricing and availability is not currently available.