Merck
  • A Caco-2 cell based screening method for compounds interacting with MRP2 efflux protein.
A Caco-2 cell based screening method for compounds interacting with MRP2 efflux protein.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2008-09-09)
Sanna Siissalo, Jenni Hannukainen, Johanna Kolehmainen, Jouni Hirvonen, Ann Marie Kaukonen
ABSTRACT
The aim of this work was to develop a screening method for MRP2 efflux substrates using the well-characterized, human-based intestinal Caco-2 cell model as a platform. MRP2 has a significant role in drug absorption and disposition and is known to co-operate with phase II metabolic enzymes. Caco-2 cells grown in a 96-well plate were loaded with non-fluorescent CDCFDA (diacetate ester of 5(6)-carboxy-2',7'-dichlorofluorescein), which is hydrolyzed to fluorescent CDCF by intracellular esterases. De-esterification in Caco-2 was comparable to that in porcine liver esterases. CDCFDA enters the cells passively, while CDCF is effluxed out of the cells by the apically localized MRP2 and/or basolateral MRPs. The method was optimized with regard to several factors. In the concluding protocol, Caco-2 cells are grown on clear 96-well plates for 8 days. The loading conditions were optimized to 10 min incubation with 5 microM CDCFDA. The highest responses were obtained for samples taken at t=30 min. The samples were analyzed in black 96-well plates with a fluorescence plate reader. The Caco-2 based method utilizing the probe pair CDCFDA/CDCF provides a fast screening tool for MRP2 substrates and/or inhibitors, along with compounds having metabolites formed in Caco-2 that interact with MRP2.
Sigma-Aldrich
5(6)-Carboxy-2′,7′-dichlorofluorescein, BioReagent, suitable for fluorescence, ≥95% (TLC)
Pricing and availability is not currently available.