Merck
  • Activation of constitutive androstane receptor under the effect of hepatocarcinogenic aminoazo dyes in mouse and rat liver.
Activation of constitutive androstane receptor under the effect of hepatocarcinogenic aminoazo dyes in mouse and rat liver.
Bulletin of experimental biology and medicine (2008-05-07)
M Y Pakharukova, M A Smetanina, V I Kaledin, V F Kobzev, I V Romanova, T I Merkulova
ABSTRACT
Selective increase of DNA-binding activity of constitutive androstane receptor was detected in rat and mouse liver in response to aminoazo dyes exhibiting hepatocarcinogenic activity for these species (ortho-aminoazotoluene for mice and 3'-methyl-4-dimethylaminobenzene for rats). Competition of azo dyes with 3H-5alpha-androst-16-ene-3alpha-ol (a well-known ligand of constitutive androstane receptor) for binding to liver cell cytosol proteins was studied. Ortho-aminoazotoluene and 3'-methyl-4-dimethylaminobenzene were better competitors for cytosol proteins from mouse and rat liver, respectively.
Product Description
Supelco
o-Aminoazotoluene, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Fast Garnet GBC base, 97%
Pricing and availability is not currently available.