Merck
  • Evaluation of the enantiomers of 1-octen-3-ol and 1-octyn-3-ol as attractants for mosquitoes associated with a freshwater swamp in Florida, U.S.A.
Evaluation of the enantiomers of 1-octen-3-ol and 1-octyn-3-ol as attractants for mosquitoes associated with a freshwater swamp in Florida, U.S.A.
Medical and veterinary entomology (2007-12-21)
D L Kline, S A Allan, U R Bernier, C H Welch
ABSTRACT
Field studies were conducted at wooded wetlands in Gainesville, FL, U.S.A., to assess responses of natural populations of adult mosquitoes (Diptera: Culicidae) to American Biophysics MM-X and Coleman MD-2500 traps baited with enantiomers of 1-octen-3-ol, a naturally occurring compound, and 1-octyn-3-ol, a closely related synthetic compound. Overall, the same species of mosquitoes were attracted by all enantiomers, although the (R)-(+) isomer of octenol generally attracted more species, and it is the isomer produced in greatest proportion in nature. Traps baited with the R-enantiomer caught greater numbers of mosquitoes than those baited with the S-enantiomer of each compound, whereas traps baited with S-enantiomers were equally or slightly less attractive than those baited with carbon dioxide only.
Sigma-Aldrich
1-Octyne, 97%