Merck
  • Enantioseparation of 1-phenyl-1-propanol on cellulose-derived chiral stationary phase by supercritical fluid chromatography II. Non-linear isotherm.
Enantioseparation of 1-phenyl-1-propanol on cellulose-derived chiral stationary phase by supercritical fluid chromatography II. Non-linear isotherm.
Journal of chromatography. A (2007-02-17)
Stefan Ottiger, Johannes Kluge, Arvind Rajendran, Marco Mazzotti
ABSTRACT
The separation of the enantiomers of 1-phenyl-1-propanol by supercritical fluid chromatography on a chiral stationary phase, which consists of cellulose tris (3,5-dimethylphenylcarbamate) coated on a silica support (Chiralcel-OD), is studied under overloaded, non-linear chromatographic conditions. Pulse experiments are performed at a temperature of 30 degrees C using supercritical CO(2) modified with methanol as a mobile phase. The parameters of the binary Langmuir adsorption isotherm are determined by the inverse method, comparing experimental and simulated peak responses. Isotherm parameters are derived for modifier concentrations between 1 and 5% (w/w) and operating pressures between 125 and 185 bar, and the dependency of the isotherm parameters, namely the Henry constant and the saturation capacity, on operating conditions is investigated.
Sigma-Aldrich
1-Phenyl-1-propanol, ≥97%
Sigma-Aldrich
1-Phenyl-1-propanol, ≥97%, FG