Merck
  • The inhibitory effect of [D-Arg1,D-Phe,D-Try7,9,Leu11] substance P on endothelin-1 binding sites in rat cardiac membranes.
The inhibitory effect of [D-Arg1,D-Phe,D-Try7,9,Leu11] substance P on endothelin-1 binding sites in rat cardiac membranes.
Biochemical and biophysical research communications (1991-08-30)
X H Gu, D J Casley, W G Nayler
ABSTRACT
The specific binding of [125I]ET-1 to rat cardiac membrane fragments was inhibited by [D-Arg1,D-Phe, D-Try7,9,Leu11] substance P [substance P(D)], a potent bombesin antagonist. This inhibitory effect required high concentrations (greater than 3X10(-6)M) of substance P(D) and was accompanied by a steep increase in non-specific binding, and not a decrease in total binding. Such results indicate that substance P(D) does not competitively inhibit the specific binding of [125I]ET-1 to rat cardiac membrane fragments.
Sigma-Aldrich
[D-Arg1, D-Phe5, D-Trp7,9, Leu11]-Substance P, ≥95% (HPLC)