Merck
  • Effect of tetrabutylphosphonium cation on the physico-chemical properties of amino-acid ionic liquids.
Effect of tetrabutylphosphonium cation on the physico-chemical properties of amino-acid ionic liquids.
Chemical communications (Cambridge, England) (2006-05-24)
Junko Kagimoto, Kenta Fukumoto, Hiroyuki Ohno
ABSTRACT
Tetraalkylphosphonium-based amino-acid ionic liquids show lower viscosities and higher decomposition temperatures (>300 degrees C) than previously reported ammonium-based amino-acid ionic liquids.
Sigma-Aldrich
Tetrabutylphosphonium bromide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetrabutylphosphonium chloride, 96%
Pricing and availability is not currently available.