Merck
  • Terminal group effects on the fluorescence spectra of europium(III) nitrate complexes with a family of amide-based 2,3-dihydroxynaphthalene derivatives.
Terminal group effects on the fluorescence spectra of europium(III) nitrate complexes with a family of amide-based 2,3-dihydroxynaphthalene derivatives.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2006-03-10)
Ke-Wei Lei, Wei-Sheng Liu
ABSTRACT
Three ligands 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-diethyl(acetamide)) (L(a)), 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-diisopropyl(acetamide)) (L(b)) and 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-dibutyl(acetamide)) (L(c)) and their europium(III) nitrate complexes were synthesized. The complexes were characterized by elemental analysis, IR, fluorescence spectroscopy and conductivity. The europium atoms are coordinated by O-atoms from CO, Ar-O-C. With the difference of the ligands, the solid fluorescent intensities of the Eu complexes vary regularly. Some factors that influencing the fluorescent intensity were discussed.
Sigma-Aldrich
2,3-Dihydroxynaphthalene, ≥98.0% (HPLC)