Merck
  • Mass-analyzed threshold ionization study of vinyl bromide cation in the first excited electronic state using vacuum-ultraviolet radiation generated by four-wave mixing in Hg.
Mass-analyzed threshold ionization study of vinyl bromide cation in the first excited electronic state using vacuum-ultraviolet radiation generated by four-wave mixing in Hg.
The Journal of chemical physics (2005-12-27)
Mina Lee, Myung Soo Kim
ABSTRACT
The vibrational spectrum of the vinyl bromide cation in the first excited electronic state A 2A' was obtained by one-photon mass-analyzed threshold ionization (MATI) spectroscopy. The use of an improved vacuum-ultraviolet radiation source based on four-wave sum frequency mixing in Hg resulted in excellent sensitivity for MATI signals. From the MATI spectrum, the ionization energy to the A 2A' state of the cation was determined to be 10.9150+/-0.0006 eV. Nearly complete vibrational assignments for the MATI peaks were possible by utilizing the vibrational frequencies and Franck-Condon factors calculated at the density-functional theory (DFT) and time-dependent DFT/B3LYP levels with the 6-311+G(df,p) basis set.
Sigma-Aldrich
Vinyl bromide, 98%
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF