Merck
  • Comparison of chloroxylenol 0.5% plus salicylic acid 2% cream and benzoyl peroxide 5% gel in the treatment of acne vulgaris: a randomized double-blind study.
Comparison of chloroxylenol 0.5% plus salicylic acid 2% cream and benzoyl peroxide 5% gel in the treatment of acne vulgaris: a randomized double-blind study.
Drugs under experimental and clinical research (2004-01-08)
F Boutli, M Zioga, T Koussidou, D Ioannides, O Mourellou
ABSTRACT
A 12-week double-blind randomized study was performed to compare benzoyl peroxide 5% (BP) gel and chloroxylenol 0.5% plus salicylic acid 2% (PCMX + SA) cream (Nisal cream) for efficacy and adverse reactions. Thirty-seven volunteers participated in the study, 19 in the BP group and 18 in the PCMX + SA group. The patients applied the medication twice daily to the entire face. Clinical evaluation and lesion counts were obtained at 0, 3, 6, 9 and 12 weeks. At week 12 both groups showed a marked improvement in both inflammatory and noninflammatory lesions (60% and 54% for the BP group and 62% and 56% for and 56% for the PCMX + SA group, respectively). Although PCMX + SA showed a slightly stronger keratolytic effect throughout the study period, there was no statistically significant difference in the reduction of the papulopustules or comedones between the two groups. Adverse effects such as erythema and photosensitivity were significantly fewer in the PCMX + SA group at week 12 (p = 0.0002 and p = 0.05, respectively). These results suggest that PCMX + SA cream is as effective as BP gel in the treatment of papulopustular and comedonal acne and that it is better tolerated.
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, purum, ≥98.0% (T)
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, 99%