Merck
  • Understanding the radiolabelling mechanism of 99mTc-antimony sulphide colloid.
Understanding the radiolabelling mechanism of 99mTc-antimony sulphide colloid.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2003-11-19)
Chris Tsopelas
ABSTRACT
The chemistry of antimony trisulphide colloid (ATC) was examined to elucidate the radiolabelling mechanism with 99mTcO4(-). Ion exchange chromatography and atomic absorption spectrophotometry techniques determined ATC to be resistant to hydrolysis in 0.1M hydrochloric acid (HCl) at 25 degrees C or 100 degrees C (>97% recovery, Sb3+ absent). Hydrogen sulphide gas detected did not participate in the mechanism, where antimony trisulphide and 99mTcO4(-) in HCl/100 degrees C yielded 96% 99mTc-product from a K2S-free formulation (versus 98% when K2S was present). 99mTcO4(-) was reduced >90% by DMSA or dithiothreitol under the same conditions, identifying involvement of thiol groups. Infrared analysis of Re-ATC showed S=O bonds, indicating excess thiol groups at the colloid surface were oxidised at the expense of 99mTcO4(-) reduction.
Sigma-Aldrich
Antimony(III) sulfide, 99.995% trace metals basis
Sigma-Aldrich
Antimony(III) sulfide, powder