Merck
New stable backbone linker resins for solid-phase peptide synthesis.
Organic letters (2003-02-14)
Wenxin Gu, Richard B Silverman
ABSTRACT
[reaction: see text] Two new 4-methoxybenzaldehyde backbone linker resins were developed for the solid-phase synthesis of peptides. The linkers are very stable during the cleavage of common protecting groups for amines (Fmoc, Boc) and carboxylic acids (Me, All, tBu) in peptide synthesis. Cleavage from the resin with refluxing TFA is sufficiently mild for peptides containing polar and nonpolar amino acids.
Sigma-Aldrich
p-Anisaldehyde, 98%
Pricing and availability is not currently available.
Supelco
p-Anisaldehyde, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Pricing and availability is not currently available.