Merck
  • Total synthesis of the alkaloids martinelline and martinellic acid via a hetero Diels-Alder multicomponent coupling reaction.
Total synthesis of the alkaloids martinelline and martinellic acid via a hetero Diels-Alder multicomponent coupling reaction.
Organic letters (2002-08-17)
David A Powell, Robert A Batey
ABSTRACT
[reaction: see text] A concise synthesis of the guanidine alkaloids, (+/-)-martinelline and (+/-)-martinellic acid, using a protic acid catalyzed 2:1 hetero Diels-Alder coupling reaction between N-Cbz 2-pyrroline and methyl 4-aminobenzoate, is described. Protic acid catalysis, rather than Lewis acid catalysis, was necessary to achieve the desired sense of diastereocontrol in the coupling reaction.
Sigma-Aldrich
Methyl 4-aminobenzoate, 98%
Pricing and availability is not currently available.