Merck
  • Biotransformation of gamma-terpinene and (-)-alpha-phellandrene by the larvae of common cutworm (Spodoptera litura).
Biotransformation of gamma-terpinene and (-)-alpha-phellandrene by the larvae of common cutworm (Spodoptera litura).
Journal of agricultural and food chemistry (2000-07-18)
M Miyazawa, T Wada
ABSTRACT
gamma-Terpinene was mixed in artificial diet at a concentration of 1 mg/g of diet, and the diet was fed to the last instar larvae of common cutworm (Spodoptera litura). Metabolites were recovered from frass and analyzed spectroscopically. gamma-Terpinene was transformed mainly to p-mentha-1,4-dien-7-oic acid and p-cymen-7-oic acid (cumic acid). Similarly, (-)-alpha-phellandrene was transformed mainly to (4R)-p-mentha-1,5-dien-7-oic acid and p-cymen-7-oic acid (cumic acid). The C-7 position (allylic methyl group) of gamma-terpinene and (-)-alpha-phellandrene was preferentially oxidized.
Sigma-Aldrich
α-Phellandrene, ≥75%, stabilized
Pricing and availability is not currently available.
Sigma-Aldrich
(R)-(−)-α-Phellandrene, ≥95.0% (sum of enantiomers, GC)
Pricing and availability is not currently available.