Merck
  • Reductive cleavage of demeton-S-methyl by Corynebacterium glutamicum in cometabolism on more readily metabolizable substrates.
Reductive cleavage of demeton-S-methyl by Corynebacterium glutamicum in cometabolism on more readily metabolizable substrates.
Applied and environmental microbiology (2000-03-04)
L Girbal, D Hilaire, S Leduc, L Delery, J L Rols, N D Lindley
ABSTRACT
Corynebacterium glutamicum is able to biotransform demeton-S-methyl, an organophosphorus compound, during cometabolism with more readily metabolizable substrates. Among the cosubstrates used, fructose is the growth substrate that is most favorable for demeton-S-methyl biotransformation. The reaction mechanism of demeton-S-methyl biotransformation involves reductive cleavage of an S-C bond, which leads to accumulation of dimethyl thiophosphate in the culture medium.
Product Description
Supelco
Demeton-S-methyl solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard