Merck
Enhanced decomposition of NO on the alkalized PdO/Al2O3 catalyst.
Chemosphere (1999-11-27)
Y J Huang, H P Wang, C T Yeh, C C Tai, C Y Peng
ABSTRACT
Experimentally, decomposition of NO on the alkalized Pd/Al2O3 catalyst is remarkably enhanced at 825-1000 K. The enhancement in N2 yield may be due to the additional basic sites on the alkalized catalyst that can trap NO molecules. However, at T > 1000 K, due to the fact that the absorbed oxygen in subsurface or bulk of Pd was involved in the formation and desorption of oxygen molecules, yield of oxygen was enhanced.
Sigma-Aldrich
Palladium(II) oxide, 99.97% trace metals basis