Merck
  • The diagnostic significance of an assay for 'total' hepatitis C core antigen.
The diagnostic significance of an assay for 'total' hepatitis C core antigen.
Journal of virological methods (2001-07-11)
J B Kurtz, E Boxall, N Qusir, J Shirley, D Coleman, C Chandler
ABSTRACT
A measurable serological response to hepatitis C infection is delayed on average until 70 days after infection. In addition, it may not occur in some immunocompromised people. Detection of free hepatitis C (HCV) core antigen in blood has enabled diagnosis in the pre-seroconversion period. The ability to detect 'total' HCV core antigen, both free and antibody bound, would widen its use for confirming anti-HCV antibody positive patients and monitoring a therapeutic response. This study has evaluated a prototype 'total' HCV core antigen immunoassay. Sera from 145 HCV negative blood donors gave a mean value of 54.9 (+/-46.2) pg/ml based on recombinant antigen standards. Using these figures, the HCV core antigen cut-off was set as 200 pg/ml. Two hundred blood donors sera with indeterminant (a single-band on recombinant immunoblot assay) HCV antibody statuses gave fully concordant HCV core antigen results compared to their polymerase chain reactions (PCRs)--three positive, and 197 negative. HCV core antigen and PCR results were compared for 59 sera from 19 HCV positive liver disease patients. The HCV core antigen results were in complete agreement with their PCRs for the nine patients always PCR positive and the three continuously negative. For six patients on antiviral therapy whose qualitative PCRs changed from positive to negative, the HCV core antigen results paralleled the PCR results. The only discrepant results were from one patient whose PCR results went from negative to positive. 'Total' HCV core antigen testing will greatly improve the scope of diagnostic tests for hepatitis C.
Sigma-Aldrich
HBcAg core human, ~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli