Merck
  • Effects of peripheral CCK receptor blockade on gastric emptying in rats.
Effects of peripheral CCK receptor blockade on gastric emptying in rats.
American journal of physiology. Regulatory, integrative and comparative physiology (2002-10-22)
Roger D Reidelberger, Linda Kelsey, Dean Heimann, Martin Hulce
ABSTRACT
Type A CCK receptor (CCKAR) antagonists differing in blood-brain barrier permeability [devazepide penetrates; the dicyclohexylammonium salt of Nalpha-3-quinolinoyl-d-Glu-N,N-dipentylamide (A-70104) does not] were used to test the hypothesis that duodenal nutrient-induced inhibition of gastric emptying is mediated by CCKARs located peripheral to the blood-brain barrier. Rats received A-70104 (700 or 3,000 nmol. kg(-1). h(-1) iv) or devazepide (2.5 micromol/kg iv) and either a 15-min intravenous infusion of CCK-8 (3 nmol. kg(-1). h(-1)) or duodenal infusion of casein, peptone, Intralipid, or maltose. Gastric emptying of saline was measured during the last 5 min of each infusion. A-70104 and devazepide abolished the gastric emptying response to a maximal inhibitory dose of CCK-8. Each of the macronutrients inhibited gastric emptying. A-70104 and devazepide attenuated inhibitory responses to each macronutrient. Intravenous injection of a CCK antibody to immunoneutralize circulating CCK had no effect on peptone or Intralipid-induced responses. Thus endogenous CCK appears to act in part by a paracrine or neurocrine mechanism at CCKARs peripheral to the blood-brain barrier to inhibit gastric emptying.
Sigma-Aldrich
EZMix Tryptone, Pancreatic digest of casein