Merck
  • Avidity confers FcγR binding and immune effector function to aglycosylated immunoglobulin G1.
Avidity confers FcγR binding and immune effector function to aglycosylated immunoglobulin G1.
Journal of molecular recognition : JMR (2012-03-13)
Thomas C Nesspor, T Shantha Raju, Chen-Ni Chin, Omid Vafa, Randall J Brezski
ABSTRACT
Immunoglobulin G (IgG) antibodies are an integral part of the adaptive immune response that provide a direct link between humoral and cellular components of the immune system. Insights into relationships between the structure and function of human IgGs have prompted molecular engineering efforts to enhance or eliminate specific properties, such as Fc-mediated immune effector functions. Human IgGs have an N-glycosylation site at Asn297, located in the second heavy chain constant region (CH2). The composition of the Fc glycan can have substantial impacts on Fc gamma receptor(FcγR) binding. The removal of the glycan through enzymatic deglycosylation or mutagenesis of the N-linked glycosylation site has been reported to "silence" FcγR-binding and effector functions, particularly with assays that measure monomeric binding. However, interactions between IgGs and FcγRs are not limited to monomeric interactions but can be influenced by avidity, which takes into account the sum of multimeric interactions between antigen-engaged IgGs and FcγRs. We show here that under in vitro conditions, which allowed avidity binding, aglycosylated IgGs can bind to one of the FcγRs, FcγRI, and mediate effector functions. These studies highlight how the valency of a molecular interaction (monomeric binding versus avidity binding) can influence antibody/FcγR interactions such that avidity effects can translate very low intrinsic affinities into significant functional outcomes.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit