Merck
  • Structural insights of the MenD from Escherichia coli reveal ThDP affinity.
Structural insights of the MenD from Escherichia coli reveal ThDP affinity.
Biochemical and biophysical research communications (2009-04-03)
Amit Priyadarshi, Yasar Saleem, Ki Hyun Nam, Key-Sun Kim, Sam-Yong Park, Eunice EunKyeong Kim, Kwang Yeon Hwang
ABSTRACT
MenD (2-succinyl-5-enolpyruvyl-6-hydroxy-3-cyclohexadiene-1-carboxylate) synthase belongs to the superfamily of thiamin diphosphate-dependent decarboxylases, which converts isochorismate and 2-oxoglutarate to SHCHC, pyruvate, and carbon dioxide. Here, we report the first crystal structure of apo-MenD from Escherichia coli determined in tetragonal crystal form. The subunit displays the typical three-domain structure observed for ThDP-dependent enzymes. Analytical gel filtration shows that EcMenD behaves as a dimer as well as a tetramer. Circular dichroism and isothermal calorimetry results confirm EcMenD dependency on ThDP, which concomitantly helps to stabilize with better configuration.
Sigma-Aldrich
Pyruvate Oxidase from Aerococcus sp., lyophilized powder, ≥35 units/mg protein (biuret)
Sigma-Aldrich
Pyruvate Oxidase from microorganisms, lyophilized powder, ≥1.5 U/mg