MilliporeSigma
  • Immunochemical localization of keratan sulfate proteoglycans in cornea, sclera, and limbus using a keratanase-generated neoepitope monoclonal antibody.
Immunochemical localization of keratan sulfate proteoglycans in cornea, sclera, and limbus using a keratanase-generated neoepitope monoclonal antibody.
Investigative ophthalmology & visual science (2008-06-03)
Saeed Akhtar, Briedgeen C Kerr, Anthony J Hayes, Clare E Hughes, Keith M Meek, Bruce Caterson
ABSTRACT
To evaluate the use of neoepitope monoclonal antibody BKS-1, which recognizes keratanase-generated keratan sulfate (KS) stubs on keratan sulfate proteoglycans in human cornea, limbus, and sclera. BKS-1 specifically recognizes a keratanase-generated neoepitope [N-acetyl-glucosamine-6-sulfate (GlcNAc-6-S)] at the nonreducing terminal of corneal and skeletal KS glycosaminoglycan chains. It was produced by using keratanase-digested KS peptides from bovine cartilage aggrecan as the immunizing antigen. BKS-1 was used in conjunction with 5D4 to analyze the KS distribution in human cornea, limbus, and sclera using Western blotting, immunohistochemistry, and electron microscopy. 5D4 Western blot analysis displayed a diffuse staining pattern, and it was difficult to distinguish differences among cornea, sclera, and limbus. However, BKS-1 showed differences in KS levels, with higher levels in the cornea and lower levels in the limbus and sclera. Ultrastructural studies showed that the monoclonal antibody (mAb) BKS-1 neoepitope was not observed in the epithelium or basement membrane; however, 5D4 was present in these layers. Large quantities of both antibodies were present in Bowman's layer, stroma, and Descemet's membrane, but the quantity of 5D4 was significantly higher (P < 0.001) than the quantity of BKS-1 in all these layers of the cornea. mAb 5D4 recognizes oversulfated structures within KS chains, whereas BKS-1 recognizes a single neoepitope on KS after keratanase digestion of monosulfated KS disaccharides. With the use of BKS-1, the authors identified a more clearly defined pattern for KS distribution in the cornea than was seen with 5D4. The presence of a large quantity of BKS-1 immunostaining in the cornea suggests that KS-substituted proteoglycans are more prevalent in the cornea than in the limbus or sclera.