Merck
  • Cytotoxic responses to aromatic ring and configurational variations in alpha-conidendrin, podophyllotoxin, and sikkimotoxin derivatives.
Cytotoxic responses to aromatic ring and configurational variations in alpha-conidendrin, podophyllotoxin, and sikkimotoxin derivatives.
Journal of medicinal chemistry (2001-02-15)
A Dantzig, R T LaLonde, F Ramdayal, R L Shepard, K Yanai, M Zhang
ABSTRACT
Derivatives of alpha-conidendrin, podophyllotoxin, and sikkimotoxin were prepared to evaluate the cytotoxic contributions of C-4 configuration and pendant and fused arene substitutions. Dimethyl-alpha-conidendryl alcohol (5), 9-deoxypodophyllol (6), and 9-deoxysikkimol (17) were dehydrated to their respective oxolane derivatives 4, 3, and 9. Diols 5 and 6 were converted via oxabicyclo[3.2.1]octanols 10 and 14 to target oxolanes 8 and 7 where C-4 had been inverted relative to that in 3 and 4. Cytotoxicities of the five oxolanes were determined in two drug-sensitive human leukemia and two multidrug-resistant cell lines expressing P-glycoprotein or multidrug-resistance associated protein (MRP). Changing the pendant arene configuration or replacing a m-methoxy by hydrogen resulted in a 100-fold cytotoxicity loss. Replacing a methylenedioxy group in the fused arene by two methoxy substituents reduced cytotoxicity by 10-fold. Drug-resistant cell lines were equally resistant to compounds 3, 4, 8, and 9 indicating that these four compounds do not serve as substrates of the transport proteins P-glycoprotein and MRP.