Merck
  • The lack of Tex44 causes severe subfertility with flagellar abnormalities in male mice.
The lack of Tex44 causes severe subfertility with flagellar abnormalities in male mice.
Cellular & molecular biology letters (2024-05-16)
Sophie Dupuis, Marie-Sophie Girault, Morgane Le Beulze, Côme Ialy-Radio, Luis Bermúdez-Guzmán, Ahmed Ziyyat, Sandrine Barbaux
ABSTRACT
By analyzing a mouse Interspecific Recombinant Congenic Strain (IRCS), we previously identified a quantitative trait locus (QTL), called Mafq1 on mouse chromosome 1, that is associated with male hypofertility and ultrastructural sperm abnormalities. Within this locus, we identified a new candidate gene that could be implicated in a reproductive phenotype: Tex44 (Testis-expressed protein 44). We thus performed a CRISPR/Cas9-mediated complete deletion of this gene in mice in order to study its function. Tex44-KO males were severely hypofertile in vivo and in vitro due to a drastic reduction of sperm motility which itself resulted from important morphological sperm abnormalities. Namely, Tex44-KO sperm showed a disorganized junction between the midpiece and the principal piece of the flagellum, leading to a 180° flagellar bending in this region. In addition, the loss of some axonemal microtubule doublets and outer dense fibers in the flagellum's principal piece has been observed. Our results suggest that, in mice, TEX44 is implicated in the correct set-up of the sperm flagellum during spermiogenesis and its absence leads to flagellar abnormalities and consequently to severe male hypofertility.
Sigma-Aldrich
Anti-SEPT4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-TEX44 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution