Merck
  • mGlu3 Metabotropic Glutamate Receptors as a Target for the Treatment of Absence Epilepsy: Preclinical and Human Genetics Data.
mGlu3 Metabotropic Glutamate Receptors as a Target for the Treatment of Absence Epilepsy: Preclinical and Human Genetics Data.
Current neuropharmacology (2022-05-18)
Roberta Celli, Pasquale Striano, Rita Citraro, Luisa Di Menna, Milena Cannella, Tiziana Imbriglio, Mahmoud Koko, Euro Epinomics-Cogie Consortium, Giovambattista De Sarro, James A Monn, Giuseppe Battaglia, Gilles Van Luijtelaar, Ferdinando Nicoletti, Emilio Russo, Antonio Leo
ABSTRACT
Previous studies suggest that different metabotropic glutamate (mGlu) receptor subtypes are potential drug targets for treating absence epilepsy. However, no information is available on mGlu3 receptors. To examine whether (i) changes of mGlu3 receptor expression/signaling are found in the somatosensory cortex and thalamus of WAG/Rij rats developing spontaneous absence seizures; (ii) selective activation of mGlu3 receptors with LY2794193 affects the number and duration of spikewave discharges (SWDs) in WAG/Rij rats; and (iii) a genetic variant of GRM3 (encoding the mGlu3 receptor) is associated with absence epilepsy. Animals: immunoblot analysis of mGlu3 receptors, GAT-1, GLAST, and GLT-1; realtime PCR analysis of mGlu3 mRNA levels; assessment of mGlu3 receptor signaling; EEG analysis of SWDs; assessment of depressive-like behavior. Humans: search for GRM3 and GRM5 missense variants in 196 patients with absence epilepsy or other Idiopathic Generalized Epilepsy (IGE)/ Genetic Generalized Epilepsy (GGE) and 125,748 controls. mGlu3 protein levels and mGlu3-mediated inhibition of cAMP formation were reduced in the thalamus and somatosensory cortex of pre-symptomatic (25-27 days old) and symptomatic (6-7 months old) WAG/Rij rats compared to age-matched controls. Treatment with LY2794193 (1 or 10 mg/kg, i.p.) reduced absence seizures and depressive-like behavior in WAG/Rij rats. LY2794193 also enhanced GAT1, GLAST, and GLT-1 protein levels in the thalamus and somatosensory cortex. GRM3 and GRM5 gene variants did not differ between epileptic patients and controls. We suggest that mGlu3 receptors modulate the activity of the cortico-thalamo-cortical circuit underlying SWDs and that selective mGlu3 receptor agonists are promising candidate drugs for absence epilepsy treatment.
Sigma-Aldrich
Anti-GABA Transporter-1 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.