Merck
  • Ac-FLTD-CMK inhibits pyroptosis and exerts neuroprotective effect in a mice model of traumatic brain injury.
Ac-FLTD-CMK inhibits pyroptosis and exerts neuroprotective effect in a mice model of traumatic brain injury.
Neuroreport (2021-01-21)
Pengfei Wang, Baogen Pan, Jun Tian, Lei Yang, Zeshang Chen, Lijun Yang, Zhenzeng Fan
ABSTRACT
Pyroptosis has been reported to contribute to the traumatic brain injury (TBI) process. Ac-FLTD-CMK is a newly synthesized pyroptosis inhibitor. However, whether Ac-FLTD-CMK inhibits pyroptosis and plays a neuroprotective role after TBI is unknown. The present study aimed to determine the effects of Ac-FLTD-CMK on TBI in a mouse model. Male C57BL/6 mice were randomly divided into sham, TBI + vehicle, and TBI + Ac-FLTD-CMK groups. TBI was induced using a weight-drop apparatus. Intraventricular injection of Ac-FLTD-CMK was performed 30 min after TBI. Caspase-1, caspase-11, gasdermin-D (GSDMD), and caspase-3 expression in the peri-contusional cortex were assessed by western blotting. Interleukin-1β (IL-1β) and interleukin-18 (IL-18) expression in the peri-contusional cortex were measured using ELISA. Behavioral experiments, brain water content, Evans blue extravasation, lactate dehydrogenase (LDH) release, and terminal deoxynucleotidyl transferase-mediated dUTP-biotin nick end labeling staining were also performed. The results showed that Ac-FLTD-CMK administration significantly downregulated caspase-1 p20, caspase-11 p20, GSDMD N-terminal, IL-1β, and IL-18 expression; reduced LDH release; alleviated neuronal death; attenuated brain edema and blood-brain barrier damage; and improved neurobehavioral function. These findings indicate that Ac-FLTD-CMK treatment suppresses pyroptosis and protects mice against TBI.