- Sodium-difluoro(oxalato)borate (NaDFOB): a new electrolyte salt for Na-ion batteries.
Sodium-difluoro(oxalato)borate (NaDFOB): a new electrolyte salt for Na-ion batteries.
Chemical communications (Cambridge, England) (2015-05-20)
Juner Chen, Zhenguo Huang, Caiyun Wang, Spencer Porter, Baofeng Wang, Wilford Lie, Hua Kun Liu
PMID25987231
ABSTRACT
A new electrolyte salt, sodium-difluoro(oxalato)borate (NaDFOB), was synthesized and studied, which enables excellent reversible capacity and high rate capability when used in Na/Na0.44MnO2 half cells. NaDFOB has excellent compatibility with various common solvents used in Na-ion batteries, in strong contrast to the solvent dependent performances of NaClO4 and NaPF6. In addition, NaDFOB possesses good stability and generates no toxic or dangerous products when exposed to air and water. All these properties demonstrate that NaDFOB could be used to prepare high performance electrolytes for emerging Na-ion batteries.