Merck
  • Chemical and Instrumental Approaches for Capillary Electrophoresis (CE)-Fluorescence Analysis of Proteins.
Chemical and Instrumental Approaches for Capillary Electrophoresis (CE)-Fluorescence Analysis of Proteins.
Methods in molecular biology (Clifton, N.J.) (2016-07-31)
Isabelle Le Potier, Audrey Boutonnet, Vincent Ecochard, François Couderc
ABSTRACT
Capillary electrophoresis (CE) coupled to fluorescence detection is an invaluable technique for the quantitative analysis of proteins of interest in the field of clinical diagnosis and quality control of novel biotechnology products. The various chemical and instrumental approaches that have been reported to carry out such sensitive analysis are described in this paper. To illustrate the contribution of CE to the analysis of therapeutic proteins, a detailed protocol for impurities profiling of a recombinant antibody sample using CE-LEDIF is given.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), BioReagent, For use as a marker in SDS-PAGE