Merck
Amorphous Aggregation of Amyloid Beta 1-40 Peptide in Confined Space.
Chemphyschem : a European journal of chemical physics and physical chemistry (2015-09-06)
Giulia Foschi, Cristiano Albonetti, Fabiola Liscio, Silvia Milita, Pierpaolo Greco, Fabio Biscarini
ABSTRACT
The amorphous aggregation of Aβ1-40 peptide is addressed by using micromolding in capillaries. Both the morphology and the size of the aggregates are modulated by changing the contact angle of the sub-micrometric channel walls. Upon decreasing the hydrophilicity of the channels, the aggregates change their morphology from small aligned drops to discontinuous lines, thereby keeping their amorphous structure. Aβ1-40 fibrils are observed at high contact angles.
Sigma-Aldrich
Amyloid β Protein Fragment 1-40, ≥90% (HPLC), powder