Merck
  • Black pepper and piperine induce anticancer effects on leukemia cell line.
Black pepper and piperine induce anticancer effects on leukemia cell line.
Toxicology research (2021-04-23)
Somesh Banerjee, Parul Katiyar, Vijay Kumar, Shashank Sagar Saini, Ritu Varshney, Vengadesan Krishnan, Debabrata Sircar, Partha Roy
ABSTRACT
The black pepper, most commonly used in Indian cuisines for ages, is considered as "king of spices." The present study evaluates the anticancer potential of black pepper and its main constituent, i.e. alkaloid piperine, against human leukemia cell line, K-562 cells. Gas chromatography-mass spectrometry (GC-MS) analysis confirmed the presence of piperine in black pepper extract. The methanolic extract of black pepper (BP-M) and pure piperine (PIP) showed a strong cytotoxic effect against this cell line. Both BP-M and PIP generated apoptotic bodies in K-562 cells and caused nuclear condensation as visualized by fluorescent microscopy, which was further confirmed by flow cytometry analysis. BP-M and PIP also generated reactive oxygen species in K-562 cells as established by flow cytometry. The translation of Bax, caspase-3 and caspase-9 genes was found to be upregulated with subsequent downregulation of Bcl-2 gene. The anti-proliferative effect of both BP-M and PIP was also observed by trypan blue staining and was further confirmed by the downregulated expression of proliferating cell nuclear antigen (PCNA). The molecular docking studies showed the binding of PIP with PCNA and Bcl-2 and supported the in vitro findings. The docking studies also proposed the binding of PIP to ADP binding pocket of Apaf-1 protein. Taken together, these findings signify the anticancer potential of both black pepper and PIP, thus proposing black pepper as a potent nutraceutical for preventing the progression of chronic myeloid leukemia.
Product Description
Millipore
BCA Protein Assay Kit, The BCA protein assay is a simple and reliable protein quantification method.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid