Merck
  • The effects of genetic deletion of Macrophage migration inhibitory factor on the chronically hypoxic pulmonary circulation.
The effects of genetic deletion of Macrophage migration inhibitory factor on the chronically hypoxic pulmonary circulation.
Pulmonary circulation (2021-01-16)
Lili Li, Maojia Xu, Simon C Rowan, Katherine Howell, Adam Russell-Hallinan, Seamas C Donnelly, Paul McLoughlin, John A Baugh
ABSTRACT
While it is well established that the haemodynamic cause of hypoxic pulmonary hypertension is increased pulmonary vascular resistance, the molecular pathogenesis of the increased resistance remains incompletely understood. Macrophage migration inhibitory factor is a pleiotropic cytokine with endogenous tautomerase enzymatic activity as well as both intracellular and extracellular signalling functions. In several diseases, macrophage migration inhibitory factor has pro-inflammatory roles that are dependent upon signalling through the cell surface receptors CD74, CXCR2 and CXCR4. Macrophage migration inhibitory factor expression is increased in animal models of hypoxic pulmonary hypertension and macrophage migration inhibitory factor tautomerase inhibitors, which block some of the functions of macrophage migration inhibitory factor, and have been shown to attenuate hypoxic pulmonary hypertension in mice and monocrotaline-induced pulmonary hypertension in rats. However, because of the multiple pathways through which it acts, the integrated actions of macrophage migration inhibitory factor during the development of hypoxic pulmonary hypertension were unclear. We report here that isolated lungs from adult macrophage migration inhibitory factor knockout (MIF-/- ) mice maintained in normoxic conditions showed greater acute hypoxic vasoconstriction than the lungs of wild type mice (MIF+/+ ). Following exposure to hypoxia for three weeks, isolated lungs from MIF-/- mice had significantly higher pulmonary vascular resistance than those from MIF+/+ mice. The major mechanism underlying the greater increase in pulmonary vascular resistance in the hypoxic MIF-/- mice was reduction of the pulmonary vascular bed due to an impairment of the normal hypoxia-induced expansion of the alveolar capillary network. Taken together, these results demonstrate that macrophage migration inhibitory factor plays a central role in the development of the pulmonary vascular responses to chronic alveolar hypoxia.
Sigma-Aldrich
Human Pulmonary Microvascular Endothelial Cells (HPMEC), 500,000 cryopreserved cells
Pricing and availability is not currently available.