Merck
  • The effect of ginger supplementation on IL2, TNFα, and IL1β cytokines gene expression levels in patients with active rheumatoid arthritis: A randomized controlled trial.
The effect of ginger supplementation on IL2, TNFα, and IL1β cytokines gene expression levels in patients with active rheumatoid arthritis: A randomized controlled trial.
Medical journal of the Islamic Republic of Iran (2020-04-14)
Naheed Aryaeian, Mahdi Mahmoudi, Farhad Shahram, Shiva Poursani, Fatemeh Jamshidi, Hajar Tavakoli
ABSTRACT
Background: Rheumatoid arthritis (RA) is a chronic autoimmune and inflammatory disease that affects the joints and consequently leads to the destruction of cartilage and bone lesions. Traditionally, ginger has been consumed in treatment of osteoarthritis, joint and muscle pain, neurological diseases, and inflammation of gums, tooth pain, asthma, stroke, diabetes, and constipation. The aim of this study was to determine the effect of ginger on some immunological and inflammatory markers in patients with rheumatoid arthritis. Methods: In this study, which was performed during 2013-2016, 66 patients with active rheumatoid arthritis who referred to the rheumatology clinic at Shariati hospital were en-rolled. Patients were randomly divided into 2 groups: one group consumed 1.5 gr ginger per day, and the other group took roasted wheat flour (placebo), respectively. To determine the effect of confounding factors on the findings of the study, questionnaires for nutrient intake, physical activity, and medication were filled, and BMI was measured. For each participant, at the beginning and end of the study, Serum hs-CRP and mRNA levels of IL-1β, IL-2 and TNF-α were determined by ELISA and Quantitative Real Time PCR, respectively. Statistical analysis was performed using SPSS software. Significance level was set at p<0.05. Results: Results of the study showed ginger powder supplementation caused a significant decline in CRP (p=0.050) and IL-1β mRNA level (p=0.021). TNFα mRNA levels reduced in ginger group compared to placebo groupalthough the difference was not significant between the 2 groups (p=0.093). Ginger had no effects on IL2 gene expression. Conclusion: This study showed that ginger reduces inflammatory factors hs-CRP and IL-1β gene expression in patients with active RA and it seems that ginger can improve the inflam-mation in the patients.
Sigma-Aldrich
Human Mononuclear Cells: HMNC, adult