Merck
  • Functional expression and analysis of a human HLA-DQ restricted, nickel-reactive T cell receptor in mouse hybridoma cells.
Functional expression and analysis of a human HLA-DQ restricted, nickel-reactive T cell receptor in mouse hybridoma cells.
The Journal of investigative dermatology (1999-09-01)
J Vollmer, H U Weltzien, A Dormoy, F Pistoor, C Moulon
ABSTRACT
Nickel-induced contact dermatitis represents a T cell mediated delayed type hyperreactivity. The elucidation of the molecular basis of T cell activation by Ni2+ ions may serve as a model for the understanding of other metal allergies. We describe here the expression of hybrid T cell antigen receptor (TCR) alpha- and beta-genes, containing rearranged human Ni-reactive variable and mouse constant regions, together with human CD4 in a mouse T cell hybridoma. The resulting hybridoma specifically responds to IL-2 secretion to Ni, but not to other metal ions in the presence of HLA-matched antigen-presenting cells. Loss of CD4 decreases, but does not completely abrogate this reactivity. The restricting HLA-DQ element is identified as consisting of DQA1*0101 and DQB1*0501; however, only some of the B cell lines homozygous for these molecules effectively present Ni to the hybridoma. We interpret these data to show that (i) Ni-reactivity is definitely mediated by alpha beta TCR variable regions; (ii) as for peptide-specific TCR, the CD4 co-receptor enhances Ni-reactivity, but is not absolutely essential; (iii) Ni2+ ions like nominal peptide antigens require HLA (here class II) molecules of the APC for presentation; (iv) the restricting molecule may require a special conformation or the association with a particular type of peptide or an as yet unidentified other surface structure on the antigen-presenting cell for effective Ni-presentation.
Sigma-Aldrich
IgG1−FITC Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.