Merck
  • Protein Disulfide Isomerase A4 Is Involved in Genome Uncoating during Human Astrovirus Cell Entry.
Protein Disulfide Isomerase A4 Is Involved in Genome Uncoating during Human Astrovirus Cell Entry.
Viruses (2021-01-06)
Nayeli Aguilar-Hernández, Lena Meyer, Susana López, Rebecca M DuBois, Carlos F Arias
ABSTRACT
Although human astroviruses (HAstVs) are important agents of gastroenteritis in young children, the studies aimed at characterizing their biology have been limited, in particular regarding their cell entry process. It has been shown that HAstV serotype 8 enters human cells by a classical clathrin-mediated endocytosis pathway; however, the cell receptor or other cell entry factors that may be relevant for an efficient viral infection are unknown. In this work we used a far-Western blotting approach to identify cellular proteins that interact with the recombinant capsid spike proteins of HAstV serotypes 1, 2, and 8, synthesized in Escherichia coli. We identified the 72 kDa protein disulfide isomerase A4 (PDIA4) as a binding partner for HAstV-1 and -8 spikes, but not for the HAstV-2 spike. In agreement with this observation, the PDI inhibitor 16F16 strongly blocked infection by HAstV serotypes 1 and 8, but not serotype 2. RNA interference of PDIA4 expression selectively blocked HAstV-8 infectivity. We also showed that the PDI activity does not affect virus binding or internalization but is required for uncoating of the viral genome.
Sigma-Aldrich
PDI inhibitor 16F16, ≥98% (HPLC)
Pricing and availability is not currently available.