Merck
  • Thinned-skull cranial window technique for long-term imaging of the cortex in live mice.
Thinned-skull cranial window technique for long-term imaging of the cortex in live mice.
Nature protocols (2010-02-06)
Guang Yang, Feng Pan, Christopher N Parkhurst, Jaime Grutzendler, Wen-Biao Gan
ABSTRACT
Imaging neurons, glia and vasculature in the living brain has become an important experimental tool for understanding how the brain works. Here we describe in detail a protocol for imaging cortical structures at high optical resolution through a thinned-skull cranial window in live mice using two-photon laser scanning microscopy (TPLSM). Surgery can be performed within 30-45 min and images can be acquired immediately thereafter. The procedure can be repeated multiple times allowing longitudinal imaging of the cortex over intervals ranging from days to years. Imaging through a thinned-skull cranial window avoids exposure of the meninges and the cortex, thus providing a minimally invasive approach for studying structural and functional changes of cells under normal and pathological conditions in the living brain.
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%