Merck
  • Superresolution Microscopy of Drosophila Indirect Flight Muscle Sarcomeres.
Superresolution Microscopy of Drosophila Indirect Flight Muscle Sarcomeres.
Bio-protocol (2021-03-05)
Szilárd Szikora, Tibor Novák, Tamás Gajdos, Miklós Erdélyi, József Mihály
ABSTRACT
Sarcomeres are extremely highly ordered macromolecular assemblies where proper structural organization is an absolute prerequisite to the functionality of these contractile units. Despite the wealth of information collected, the exact spatial arrangement of many of the H-zone and Z-disk proteins remained unknown. Recently, we developed a powerful nanoscopic approach to localize the sarcomeric protein components with a resolution well below the diffraction limit. The ease of sample preparation and the near crystalline structure of the Drosophila flight muscle sarcomeres make them ideally suitable for single molecule localization microscopy and structure averaging. Our approach allowed us to determine the position of dozens of H-zone and Z-disk proteins with a quasi-molecular, ~5-10 nm localization precision. The protocol described below provides an easy and reproducible method to prepare individual myofibrils for dSTORM imaging. In addition, it includes an in-depth description of a custom made and freely available software toolbox to process and quantitatively analyze the raw localization data.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Cysteamine hydrochloride, ≥98% (titration)