Merck
  • Rosiglitazone attenuates high glucose-induced proliferation, inflammation, oxidative stress and extracellular matrix accumulation in mouse mesangial cells through the Gm26917/miR-185-5p pathway.
Rosiglitazone attenuates high glucose-induced proliferation, inflammation, oxidative stress and extracellular matrix accumulation in mouse mesangial cells through the Gm26917/miR-185-5p pathway.
Endocrine journal (2021-04-02)
Dongbo Zhao, Junli Guo, Lingping Liu, Ying Huang
ABSTRACT
Rosiglitazone (RSG) is widely used to reduce the amount of sugar in the blood of patients with diabetes mellitus. Diabetic nephropathy is the most common microvascular complication of diabetes. The role of RSG in diabetic nephropathy is not fully understood. Diabetic nephropathy model was constructed in high glucose (HG)-treated mouse mesangial cells. The effects of RSG on cell viability and cell cycle were investigated using cell counting kit-8 (CCK-8) assay and flow cytometry assay. Oxidative stress was assessed according to ROS production and SOD activity in cells. Inflammatory responses were assessed according to the releases of inflammatory cytokines. Extracellular matrix (ECM) accumulation was determined by the levels of fibronectin and collagen IV using western blot. The expression of Gm26917 and microRNA-185-5p (miR-185-5p) was detected by quantitative real-time polymerase chain reaction (qPCR). The interaction between Gm26917 and miR-185-5p was validated by dual-luciferase reporter assay, RNA immunoprecipitation (RIP) assay and pull-down assay. RSG significantly inhibited HG-induced proliferation, oxidative stress, inflammatory responses and ECM accumulation in mouse mesangial cells. The expression of Gm26917 was induced by HG but weakened by RSG. Gm26917 knockdown alleviated HG-induced proliferation, oxidative stress, inflammatory responses and ECM accumulation in mouse mesangial cells, and Gm26917 overexpression partly abolished the effects of RSG. Moreover, miR-185-5p was a target of Gm26917, and miR-185-5p inhibition recovered proliferation, oxidative stress, inflammatory responses and ECM accumulation in mouse mesangial cells that were alleviated by Gm26917 knockdown. RSG ameliorated HG-induced mouse mesangial cell proliferation, oxidative stress, inflammation and ECM accumulation partially by governing the Gm26917/miR-185-5p pathway.
Sigma-Aldrich
Anti-Collagen IV antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.