Merck
Identification of some new minor acids from chicken skin lipids.
Poultry science (1978-05-01)
R J Horvat
PMID674060
ABSTRACT
Lipids from the skin of commercially processed broilers were extracted by Folch's technique and analyzed for fatty acids as their methyl esters by gas-liquid chromatography/mass pectrometry. Of the 24 fatty acids, identified, 5 had not been previously identified in either total chicken skin lipids or skin lipid fractions. They are: pentadecenoic acid (15:1), heptadecenoic acid (17:1), arachidic acid (20:0), heneicosanoic acid (21:0), and erucic acid (22:1).
Sigma-Aldrich
Methyl heneicosanoate, ~99% (capillary GC), crystalline