Merck
  • Homocitrate is a component of the iron-molybdenum cofactor of nitrogenase.
Homocitrate is a component of the iron-molybdenum cofactor of nitrogenase.
Biochemistry (1989-04-04)
T R Hoover, J Imperial, P W Ludden, V K Shah
ABSTRACT
When apodinitrogenase (lacking FeMo-co) was activated with FeMo-co synthesized in vitro in the presence of 3H-labeled homocitrate, label was incorporated into dinitrogenase. The physical association of the label with FeMo-co was demonstrated by reisolation and purification of the cofactor from dinitrogenase. The presence of homocitrate in FeMo-co was established by NMR analysis of the organic acid extracted from dinitrogenase. Quantitation of homocitrate in dinitrogenase showed it to be present at a 1:1 ratio with molybdenum.
Sigma-Aldrich
Sodium (±)-homocitrate tribasic