Merck
  • Functional self-assembling bolaamphiphilic polydiacetylenes as colorimetric sensor scaffolds.
Functional self-assembling bolaamphiphilic polydiacetylenes as colorimetric sensor scaffolds.
Journal of the American Chemical Society (2004-07-09)
Jie Song, Justin S Cisar, Carolyn R Bertozzi
ABSTRACT
Conjugated polymers capable of responding to external stimuli by changes in optical, electrical, or electrochemical properties can be used for the construction of direct sensing devices. Polydiacetylene-based systems are attractive for sensing applications due to their colorimetric response to changes in the local environment. Here we present the design, preparation, and characterization of self-assembling functional bolaamphiphilic polydiacetylenes (BPDAs) inspired by nature's strategy for membrane stabilization. We show that by placing polar headgroups on both ends of the diacetylene lipids in a transmembranic fashion and by altering the chemical nature of the polar surface residues, the conjugated polymers can be engineered to display a range of radiation-, thermal-, and pH-induced colorimetric responses. We observed dramatic nanoscopic morphological transformations accompanying charge-induced chromatic transitions, suggesting that both side-chain disordering and main-chain rearrangement play important roles in altering the effective conjugation lengths of the poly(ene-yne). These results establish the foundation for further development of BPDA-based colorimetric sensors.