Merck
  • Exploiting self-assembled soft systems based on surfactants, biopolymers and their mixtures for inhibition of Citral degradation under harsh acidic Conditions.
Exploiting self-assembled soft systems based on surfactants, biopolymers and their mixtures for inhibition of Citral degradation under harsh acidic Conditions.
Food chemistry (2020-10-05)
Parvaiz Ahmad Bhat, Nighat Nazir, Oyais Ahmad Chat, Aijaz Ahmad Dar
ABSTRACT
The chemical instability of Citral in acidic conditions is viewed as hurdle to commercialize it in food/beverage industries. We attempted to stabilize citral in various single and mixed surfactant systems at pH 1.0 and temperature 25 °C. The study highlights the importance of amount and density of positive charge of cationic surfactants and oxyethylene content of nonionic surfactants at the interface of self-assembly in inhibiting citral degradation. The hybrid of Chitosan and P123 showed a significant increase in the half-life of citral compared to that in its individual components. The results of the study suggest that it is possible to stabilize citral in strong acidic environs having a pH as low as 1.0 using mixed surfactant or polymer-amphiphile systems with significant positive charge/number of oxyethylene in their single components. Such polymer-surfactant systems formulations if biocompatible/food grade may act as promising media to enhance shelf life of citral.
Sigma-Aldrich
Lauroyl chloride, 98%