Merck
  • The phosphatidylinositol transfer protein PITP-1 facilitates fast recovery of eating behavior after hypoxia in the nematode Caenorhabditis elegans.
The phosphatidylinositol transfer protein PITP-1 facilitates fast recovery of eating behavior after hypoxia in the nematode Caenorhabditis elegans.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-12-29)
Zohar Abergel, Maayan Shaked, Virendra Shukla, Zheng-Xing Wu, Einav Gross
ABSTRACT
Among the fascinating adaptations to limiting oxygen conditions (hypoxia) is the suppression of food intake and weight loss. In humans, this phenomenon is called high-altitude anorexia and is observed in people suffering from acute mountain syndrome. The high-altitude anorexia appears to be conserved in evolution and has been seen in species across the animal kingdom. However, the mechanism underlying the recovery of eating behavior after hypoxia is still not known. Here, we show that the phosphatidylinositol transfer protein PITP-1 is essential for the fast recovery of eating behavior after hypoxia in the nematode Caenorhabditis elegans. Unlike the neuroglobin GLB-5 that accelerates the recovery of eating behavior through its function in the oxygen (O2 )-sensing neurons, PITP-1 appears to act downstream, in neurons that express the mod-1 serotonin receptor. Indeed, pitp-1 mutants display wild-type-like O2 -evoked-calcium responses in the URX O2 -sensing neuron. Intriguingly, loss-of-function of protein kinase C 1 (PKC-1) rescues pitp-1 mutants' recovery after hypoxia. Increased diacylglycerol (DAG), which activates PKC-1, attenuates the recovery of wild-type worms. Together, these data suggest that PITP-1 enables rapid recovery of eating behavior after hypoxia by limiting DAG's availability, thereby limiting PKC activity in mod-1-expressing neurons.
Product Description
Supelco
Aldicarb, PESTANAL®, analytical standard
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder